LDS-sample buffer (4X), reducing, Thermo Scientific
Catalog # AAJ61942-AD
Supplier: Thermo Scientific Chemicals
CAS Number:
Specifications
- Size:50 mL
- Environmentally Preferable:
- Physical Form:Liquid
- Appearance:Blue
- Cat. No.:AAJ61942-AD
- Supplier no.:J61942-AD
Specifications
About this item
LDS-sample buffer (4X), reducing is used to prepare protein samples for denaturing gel electrophoresis. It is used in 1D gel electrophoresis, bis-tris gel electrophoresis, tris-acetate gel electrophoresis, protein Gel electrophoresis, proteins, expression, isolation & analysis.
Physical Form: Viscous liquid
Appearance: Blue
UN No.:UN3082
This buffer contains 988mM Tris-HCl, 2.04mM EDTA, 8% LDS, 40% glycerol, 0.88% Coomassie blue, and 0.7mM phenol red, and 8% mercaptoethanol at pH 7.5.