Order Entry
United States
ContactUsLinkComponent
 
Pig Insulin (from Pancreas)
  AAJ61321-MB
 :  Avantor
 :  
Pig Insulin (from Pancreas)
  AAJ61321-MB
 :  Avantor
 :  

 

  • CAS No.:
    12584-58-6
  • Source:
    Pancreas
  • Species:
    Pig
  • Size:
    25 mg
  • Environmentally Preferable:
  • Storage Conditions:
    –30...–10 °C
  • Form:
    Lyophilized powder
  • MDL No.:
    MFCD00131380
  • Protein/Peptide Name:
    Insulin
  • Purity:
    97%
  • Molecular Weight:
    5777.59
  • Color:
    White
  • Molecular Formula:
    C₂₅₆H₃₈₁N₆₅O₇₆S₆
  • Cat. No.:
    AAJ61321-MB
  • Supplier no.:
    J61321-MB

 

 

Applications: A hormone that is central to regulating energy and glucose metabolism in the body

Purity: 97%
CAS No.: 12584-58-6
Molecular Formula: C256H381N65O76S6
Formula Weight: 5777.59
Storage Temperature: -30°C to -10°C
Physical Form: Lyophilized powder
Appearance: White
MDL No.: MFCD00131380