Zetex Heat Resistant Tape, Newtex Industries
Catalog # 75794-942
Supplier: Newtex Industries
Specifications
- Width:1"
- Length:100'
- Environmentally Preferable:
- Cat. No.:75794-942
- Supplier no.:1200001
Specifications
About this item
Heat resistant tape withstands temperatures up to 538 °C (1000 °F).
Zetex tape can be used in a broad range of applications including gasketing, lagging, insulation, and encapsulation. Can be easily wrapped around water and steam pipes, hydraulic lines, and auto exhaust.
Manufacturer uses an overspray process to preserve texturization in fabrication and use. Zetex is made from inorganic materials, so the product will not smolder or produce hazardous outgassing. One roll of tape is 100' long. The tape is 1" wide and 0.03" thick.