To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Proteoglycan 2 Bone Marrow (natural killer cell activator eosinophil granule major basic Protein
- Antigen Symbol:PRG2
- Clonality:Monoclonal
- Clone:1.B.787
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:50 μg
- Environmentally Preferable:
- Gene ID:5553
- Antigen Synonyms:proteoglycan 2|BMPG|MBP1|MBP|bone marrow (natural killer cell activator|eosinophil granule major basic protein)
- Storage Buffer:Preservative: 10mM Sodium Azide; Constituents: 1mg/ml BSA, PBS. pH 7.2
- Storage Temperature:Store at –20 °C. Aliquots are stable for 12 months.
- Concentration:0.05 mg/ml
- Immunogen:Full length protein (Human).
- Tested Applications:ELISA, IHC-P, IHC-Fr, WB
- Purification:Protein A purified
- Cat. No.:89360-012
- Supplier no.:GTX14462
Specifications
About this item
PRG2 is a cytotoxin and helminthotoxin. Also induces noncytolytic histamine release from human basophils. Involved in antiparasitic defense mechanisms and immune hypersensitivity reactions. The proform acts as a proteinase inhibitor, reducing the activity of PAPPA. In pregnancy serum, the proform exists as a disulfide-linked 2:2 heterotetramer with PAPPA, as a disulfide-linked 2:2 heterotetramer with AGT, and as a complex (probably a 2:2:2 heterohexamer) with AGT and C3dg.
ELISA: Use at an assay dependent dilution. IHC-P: Use at an assay dependent dilution. Perform enzymatic antigen retrieval before commencing with IHC staining protocol. IHC-Fr: Use at an assay dependent dilution. WB: Use at an assay dependent dilution. Predicted molecular weight: 26.6 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: PRG2
Clonality: Monoclonal
Clone: 1.B.787
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human