Order Entry
United States
Orders LinkContactUsLinkComponent
Anti-MT PAP Rabbit Polyclonal Antibody

Anti-MT PAP Rabbit Polyclonal Antibody

100 μl
Catalog # 89358-938
Supplier:   Genetex
Supplier #:  GTX118983
Anti-MT PAP Rabbit Polyclonal Antibody
Catalog # 89358-938
Supplier:  Genetex
Anti-MT PAP Rabbit Polyclonal Antibody

100 μl

Price
$600.96
Log in for availability
Each (100µl)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

The protein encoded by this gene is a member of the DNA polymerase type-B-like family. This enzyme synthesizes the 3' poly(A) tail of mitochondrial transcripts and plays a role in replication-dependent histone mRNA degradation.

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: MT PAP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Specifications
  • Size:
    100 μl
  • Clonality:
    Polyclonal
  • Molecular Weight:
    66 kDa
  • Purification:
    Purified by antigen-affinity chromatography.
  • Storage Buffer:
    0.1M Tris, 0.1M Glycine, 20% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
  • Tested Applications:
    WB
  • Isotype:
    IgG
  • Host:
    Rabbit
  • Concentration:
    1 mg/ml
  • Cat. No.:
    89358-938
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Antibody Type:
    Primary
  • Antigen Symbol:
    MT PAP
  • Gene ID:
    55149
  • Immunogen:
    Recombinant protein encompassing a sequence within the center region of human Mt PAP. The exact sequence is proprietary.
  • Conjugation:
    Unconjugated
  • Antigen Synonyms:
    SPAX4|PAPD1
  • Antigen Name:
    Mitochondrial Poly(A) Polymerase
  • Storage Temperature:
    Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.