To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Kruppel-like Factor 10
- Antigen Symbol:KLF10
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:7071
- Antigen Synonyms:TIEG|TIEG1|EGR-alpha|EGRA
- Storage Buffer:1XPBS, 1% BSA, 20% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
- Molecular Weight:53 kDa
- Storage Temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:1mg/ml
- Immunogen:Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human KLF10/TIEG1. The exact sequence is proprietary.
- Tested Applications:WB
- Purification:Purified by antigen-affinity chromatography.
- Cat. No.:89355-650
- Supplier no.:GTX108661
Specifications
About this item
Rabbit Polyclonal antibody to KLF10/TIEG1 (Kruppel-like factor 10)
Suggested starting dilutions are as follows: WB: 1:500-1:10000, IP: 1:100-1:500. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: KLF10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human