Order Entry
United States
ContactUsLinkComponent
Anti-CEP 164 Mouse Monoclonal Antibody [clone: 17]
Anti-CEP 164 Mouse Monoclonal Antibody [clone: 17]
Catalog # 89364-662
Supplier:  Genetex
undefined
Anti-CEP 164 Mouse Monoclonal Antibody [clone: 17]
Catalog # 89364-662
Supplier:  Genetex
Supplier Number:  GTX70096
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Centrosomal Protein 164Kda
  • Antigen Symbol:
    CEP164
  • Clonality:
    Monoclonal
  • Clone:
    17
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100 μl
  • Environmentally Preferable:
  • Gene ID:
    22897
  • Antigen Synonyms:
    Cep164|NPHP15|KIAA1052|Centrosomal protein of 164 kDa
  • Storage Buffer:
    PBS
  • Storage Temperature:
    For short term use, store at 4 °C. For extended use, aliquot and store at –20 °C. Avoid repeated freeze-thaw cycles.
  • Concentration:
    1 mg/ml
  • Tested Applications:
    IP, WB
  • Purification:
    Affinity purified by Protein G.
  • Cat. No.:
    89364-662
  • Supplier no.:
    GTX70096

Specifications

About this item

Mouse monoclonal antibody [17] to Cep 164

Suggested starting dilutions are as follows: WB: 1:500-1:20000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: CEP 164
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: