Order Entry
United States
ContactUsLinkComponent
Anti-GP9 Mouse Monoclonal Antibody [clone: GRP-P]
Anti-GP9 Mouse Monoclonal Antibody [clone: GRP-P]
Catalog # 89365-578
Supplier:  Genetex
Anti-GP9 Mouse Monoclonal Antibody [clone: GRP-P]
Catalog # 89365-578
Supplier:  Genetex
Supplier Number:  GTX74779
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Glycoprotein 9
  • Antigen Symbol:
    GP9
  • Clonality:
    Monoclonal
  • Clone:
    GRP-P
  • Conjugation:
    Unconjugated
  • Flow Cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Size:
    200 test
  • Environmentally Preferable:
  • Gene ID:
    2815
  • Antigen Synonyms:
    glycoprotein 9|GP-IX|platelet glycoprotein IX|CD42a|Platelet GPIX|GPIX
  • Storage Buffer:
    Phosphate buffered saline containing 0.09% Sodium Azide (NaN3)
  • Storage Temperature:
    Store at +4 °C or at –20 °C if preferred. Storage in frost-free freezers is not recommended. This product should be stored undiluted. Should this product contain a precipitate we recommend microcentrifugation before use.
  • Immunogen:
    Human red blood cells and platelets
  • Tested Applications:
    Fluorescence-Activated Cell Sorting
  • Purification:
    Purified IgG prepared by ion exchange chromatography
  • Cat. No.:
    89365-578
  • Supplier no.:
    GTX74779

Specifications

About this item

Mouse monoclonal antibody [GRP-P] to CD42a

Recommended Starting Dilutions: Flow Cytometry: 1/10 - 1/100 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: GP9
Clonality: Monoclonal
Clone: GRP-P
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Dog, Mink