Order Entry
United States
ContactUsLinkComponent
Anti-CD4 Mouse Monoclonal Antibody [clone: W3/25]
Anti-CD4 Mouse Monoclonal Antibody [clone: W3/25]
  89366-050
 :  Genetex
Anti-CD4 Mouse Monoclonal Antibody [clone: W3/25]
  89366-050
 :  Genetex
 :  GTX76316
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Antibody Type:
    Primary
  • Antigen Name:
    CD4 molecule
  • Antigen Symbol:
    CD4
  • Clonality:
    Monoclonal
  • Clone:
    W3/25
  • Conjugation:
    Biotin
  • Flow Cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Rat
  • Size:
    100 μg
  • Environmentally Preferable:
  • Gene ID:
    24932
  • Antigen Synonyms:
    L3T4|p55|CD4mut|Ly-4|W3/25
  • Storage Buffer:
    Phosphate buffered saline pH7.4
  • Storage Temperature:
    Keep as concentrated solution. Store at 4 °C short term. For extended storage aliquot and store at –20 °C or below. Avoid freeze-thaw cycles.
  • Concentration:
    0.1 mg/ml
  • Immunogen:
    Rat thymocyte membrane
  • Tested Applications:
    Fluorescence-Activated Cell Sorting
  • Purification:
    Protein G affinity purified
  • Cat. No.:
    89366-050
  • Supplier no.:
    GTX76316

 

 

Mouse monoclonal [W3/25] to CD4 (Biotin)

  • For FACS: Use 0.5 ug to label 10^6 cells. Protocols are available upon request. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: