Order Entry
United States
ContactUsLinkComponent
Anti-CD68 Rat Monoclonal Antibody [clone: FA-11]
Anti-CD68 Rat Monoclonal Antibody [clone: FA-11]
Catalog # 89363-478
Supplier:  Genetex
Anti-CD68 Rat Monoclonal Antibody [clone: FA-11]
Catalog # 89363-478
Supplier:  Genetex
Supplier Number:  GTX41865
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    CD68 molecule
  • Antigen Symbol:
    CD68
  • Clonality:
    Monoclonal
  • Clone:
    FA-11
  • Conjugation:
    Unconjugated
  • Flow Cytometry:
    Yes
  • Host:
    Rat
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
    Mouse
  • Western Blot:
    Yes
  • Size:
    100 μg
  • Environmentally Preferable:
  • Gene ID:
    12514
  • Antigen Synonyms:
    macrosialin|CD68 antigen|GP110|scavenger receptor class D member 1|SCARD1|LAMP4|macrophage antigen CD68
  • Storage Buffer:
    Phosphate buffered saline containing 0.09% Sodium Azide
  • Storage Temperature:
    Store at +4 °C or at –20 °C if preferred. This product should be stored undiluted. Storage in frost free freezers is not recommended. Should this product contain a precipitate we recommend microcentrifugation before use.
  • Tested Applications:
    Fluorescence-Activated Cell Sorting
  • Purification:
    Purified IgG prepared by affinity chromatography on Protein G from tissue culture supernatant
  • Cat. No.:
    89363-478
  • Supplier no.:
    GTX41865

Specifications

About this item

Rat monoclonal antibody [FA-11] to CD68

Recommended Starting Dilutions: Flow Cytometry: 1/50 - 1/100 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: CD68
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: