ContactUsLinkComponent
AzureGelImagingSystems_950x790.jpg
Azure Gel Imaging Systems
Easily image DNA and protein gels images without a darkroom with the Azure 200.
Use your choice of dyes or stains and the system automatically selects the light source and filters for you - UV for Ethidium bromide-stained DNA gels, blue light for SYBR® Safe or similar dyes, white light for Silver stain or Coumassie Blue.
Product Features
  • Compact design - 38cm W X 55cm H X 36cm D, keep more space free on your bench
  • Fully upgradable - add fluorescent or chemiluminescent Western detection capabilities as your needs evolve