Wipes for delicate tasks, KIMTECH SCIENCE* KIMWIPES*
Catalog # KIMB34155
Supplier: KIMBERLY CLARK
Specifications
- Packed:1 box of 280 sheets
- Colour:White
- Description:KIMTECH SCIENCE* delicate task wipes, 1 ply
- L×W:112×213 mm
- Seq:02
- Environmentally Preferable:
- Pk:17.160
Specifications
About this item
These single ply, light-duty wipes can handle a variety of delicate tasks and easily wipe up liquid and dust.
- Anti-stat dispensing reduces lint and electrostatic discharge and controls usage
- Portability reduces waste
- One-at-a-time dispensing (pop-up dispenser)
Frequently Bought Together