Transport Boxes
Supplier: SARSTEDT
Certificates
About this item
Transport case housing is manufactured from durable polyethylene and offers optimum protection for primary receptacles during transport.
- Economical, flexible
- Central one-hand closure
- Lateral tabs for easy attachment of straps or restraints
- Transparent window label for easy identification
Transport cases are suitable for transportation of diagnostic samples and blood products.
Certifications: Complies with the requirement of P650 packaging instructions for class UN3373 of the ADR and RID.
Frequently Bought Together