Chromatography columns with Rotaflo® stopcock, Quickfit®
Supplier: SciLabware
Certificates
About this item
These chromatography columns are made from PYREX®, borosilicate glass.
- With PTFE Rotaflo® stopcock for fine control of eluent flow
- Chemically resistant glass sintered disc - porosity 0
- Ground glass socket accepts solvent reservoirs
Frequently Bought Together