Order Entry
United Kingdom
ContactUsLinkComponent
Black filter paper for the detection of light precipitates, grade MN 220
Black filter paper for the detection of light precipitates, grade MN 220
Supplier:  MACHEREY-NAGEL
Certificates
undefined
Black filter paper for the detection of light precipitates, grade MN 220
Supplier:  MACHEREY-NAGEL
Specifications
  • Catalog No:
  • MANA409018
  • MANA409005
  • MANA409009
  • MANA409011
  • MANA409012
  • MANA409015
  • MANA409007
  • Weight:
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • Thickness:
  • 0,17 mm
  • 0,17 mm
  • 0,17 mm
  • 0,17 mm
  • 0,17 mm
  • 0,17 mm
  • 0,17 mm
  • Filtration speed:
  • 45 s
  • 45 s
  • 45 s
  • 45 s
  • 45 s
  • 45 s
  • 45 s