DSS (Disuccinimidyl suberate) ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Bis(2,5-dioxopyrrolidin-1-yl) octanedioate, Suberic acid bis(N-hydroxysuccinimide ester), Disuccinimidyl octanedioate, DSS
CAS Number:68528-80-3
:
Formula:C₁₆H₂₀N₂O₈
MDL Number:MFCD00049059
Molecular Weight:368.34 g/mol
Melting Pt:166…170 °C
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together