Diethyl (2-bromoethyl)phosphonate 97%
:5324-30-1
:
:C₆H₁₄BrO₃P
:MFCD00000239
:75 °C (1 mmHg)
:1.27 g/cm³
:245.06 g/mol
:>112 °C
:Ambient
:226-193-3
:GHS07
- Environmentally Preferable:
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4590 to 1.4610 (20°C, 589 nm)
- GC:≥96.0 %
Frequently Bought Together