2-Fluoro-4-(trifluoromethyl)benzyl alcohol
Supplier: FLUOROCHEM
Certificates
About this item
Synonyms: (2-Fluoro-4-(trifluoromethyl)phenyl)methanol
CAS Number:197239-49-9
:
Formula:C₈H₆F₄O
MDL Number:MFCD00070808
Molecular Weight:194.13 g/mol
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together