Spill containment barriers, SPILLBLOCKER® Dike
Supplier: NEW PIG
Certificates
About this item
Flexible portable 100% polyurethane barrier for containment of spillages on floors.
- Barrier contains or diverts the flow of liquids, without absorbing the liquid
- Self-contained dove tail connectors allow for multiple lengths to contain larger spills
- Range of profile heights
These reusable barriers can be easily cleaned with soap and water.
Frequently Bought Together