Robotic tips for Eppendorf epMotion® automated pipetting system
Supplier: Thermo Fisher Scientific
Certificates
About this item
These automation tips for the epMotion automated pipetting system are available in non sterile, sterile, and sterile filtered versions.
- Each tip is lot-tested during production using a 15-point QC program on an epMotion workstation
- Tips have a low percentage Coefficient of Variation (% CV)
- Sterile tips are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen
Specifications
- Catalog No:
- 736-0757
- 736-0755
- 736-0758
- 736-0756
- 736-0759
- 736-0760
- 736-0762
- 736-0763
- 736-0761
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)