Lithium cobaltite (lithium cobalt(III) oxide) ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Lithium cobalt(III) oxide
CAS Number:12190-79-3
:
Formula:CoLiO₂
MDL Number:MFCD00049786
Molecular Weight:97.87 g/mol
EINECS:235-362-0
:GHS08
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together