Pycnometers, unadjusted
Pycnometer, made of borosilicate glass 3.3, Gay-Lussac type.
- With ground-in capillary stopper
- Nominal capacity printed on the bottom
- Very good chemical and temperature resistance
: DIN 12797
Frequently Bought Together
Pycnometer, made of borosilicate glass 3.3, Gay-Lussac type.