Bunsen burner, cartridge type
451-0156
- Max. temp.:1630 °C
- Description:Bunsen Usbeck
- Environmentally Preferable:
- Pk:1
Nickel plated brass Bunsen burner ideal for heating or sterilising instruments and laboratory glassware.
- With air regulation needle valve
Height: 85 mm
: Supplied without cartridge, please order cartridge separately.