4-Nitroquinoline-N-oxide, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: 4-Nitroquinoline 1-oxide, 4-Nitroquinoline N-oxide
CAS Number:56-57-5
:
Formula:C₉H₆N₂O₃
MDL Number:MFCD00006738
Molecular Weight:190.1555 g/mol
EINECS:200-281-1
UN:2811
ADR:6.1 II
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together