Potassium xanthogenate (potassium-O-ethyl dithiocarbonate), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Potassium-O-ethyldithiocarbonate, Potassium ethyl xanthate, Ethylxanthic acid potassium salt
CAS Number:140-89-6
:
Formula:C₃H₅KOS₂
MDL Number:MFCD00004931
Molecular Weight:160.30 g/mol
:M-Clarity™ quality level = MQ100
EINECS:205-439-3
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together