N-Decanoyl-N-methyl-D-glucamine (MEGA-10)
Supplier: G-Biosciences
Certificates
About this item
CAS Number:85261-20-7
:
Formula:C₁₇H₃₅NO₆
MDL Number:MFCD00036801
Molecular Weight:349.46 g/mol
Storage Temperature:Refrigerator
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together