Order Entry
United Kingdom
ContactUsLinkComponent
FITC (fluorescein-5-isothiocyanate, fluorescein isothiocyanate isomer I)
FITC (fluorescein-5-isothiocyanate, fluorescein isothiocyanate isomer I)
Catalog # PIER46424
CAS Number:  
FITC (fluorescein-5-isothiocyanate, fluorescein isothiocyanate isomer I)
Catalog # PIER46424
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047
  • Environmentally Preferable:

Specifications

About this item

Synonyms: 3',6'-Dihydroxy-5-isothiocyanato-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one, 5-FITC, Fluorescein Isothiocyanate, 5-isothiocyanatofluorescein, Fluorescein Isothiocyanate, Isomer I
 :
 :389.381 g/mol
EINECS:222-042-0
 :GHS07
Storage Temperature:Refrigerator
CAS Number:3326-32-7
Formula:C₂₁H₁₁NO₅S
MDL Number:MFCD00005063