(Ethoxycarbonylmethyl)triphenylphosphonium chloride ≥97%
Catalog # A19494.06
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Environmentally Preferable:
Specifications
About this item
Synonyms: (2-Ethoxy-2-oxoethyl)triphenylphosphonium chloride
CAS Number:17577-28-5
:
Formula:C₂₂H₂₂ClO₂P
MDL Number:MFCD00011746
:384.84 g/mol
EINECS:241-548-2
:GHS07
Melting Pt:∼ 145 °C (dec.)