Drum pumps for acids and alkalis
Supplier: BURKLE
Certificates
About this item
These drum pumps are suitable for weaker acids such as hydrochloric acid, phosphoric acid, chromic acid, battery acid, citric acid and more.
Delivery: Supplied with pump tube, PP hose connection, PVC-fabric hose, PP nozzle, PP drum adapter.
Frequently Bought Together