AEBSF hydrochloride (4-(2-aminoethyl)benzenesulphonylfluoride hydrochloride), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: 4-(2-Aminoethyl)benzenesulphonyl fluoride hydrochloride, AEBSF hydrochloride
CAS Number:30827-99-7
:
Formula:C₈H₁₁ClFNO₂S
MDL Number:MFCD00132962
Molecular Weight:239.69 g/mol
ADR:8 II
:M-Clarity™ quality level = MQ300
UN:3261
:GHS07
Storage Temperature:Refrigerator
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together