Betaine monohydrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:590-47-6
:
Formula:C₅H₁₁NO₂·1H₂O
MDL Number:MFCD00150010
Storage Temperature:Ambient
EINECS:203-490-6
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together