Order Entry
United Kingdom
ContactUsLinkComponent
Sealing tape, PTFE, Fluo-Kem®, SP Bel-Art
Sealing tape, PTFE, Fluo-Kem®, SP Bel-Art
Catalog # 817-1570
undefined
Sealing tape, PTFE, Fluo-Kem®, SP Bel-Art
Catalog # 817-1570

Specifications

  • Width:
    12,7 mm
  • Description:
    Lab thread tape, sintered
  • Environmentally Preferable:
  • Pk:
    3

Specifications

About this item

Non-sintered PTFE tape for use with desiccators, bell jars, glass joints and freeze-drying equipment for tight seal that will not seize over time.

  • Lubricates and seals irregular surfaces
  • Threads and joints without grease or adhesives

Length: 7,6 m

Colour: White

Packaging: Three per bag.