VWR®, Phosphate buffered saline (PBS) 1X, for biotechnology
Catalog # E504-500ML
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:500 mL
- Description:Phosphate buffered saline (PBS), 1X solution, pH 7,4
- Environmentally Preferable:
Specifications
About this item
137 mM sodium chloride, 2,7 mM potassium chloride, 12 mM phosphate buffer
pH 7,4