Copper(II) iodate, anhydrous ≥95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Copper iodate
CAS Number:13454-89-2
:
Formula:CuI₂O₆
MDL Number:MFCD00016055
Density:5.241
Molecular Weight:413.35 g/mol
Melting Pt:Dec.
Storage Temperature:Ambient
UN:1479
ADR:5.1 II
:GHS03
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together