Hafnium standard solution, 1000 mg/L Hf in 5% hydrochloric acid (from HfOCl?), standard for AAS, Specpure®
Molecular Formula: HfOCl₂ in 5% HCl
Physical Form: Liquid
UN No.: 1789
MDL No.: MFCD00011032
: Traceable to NIST
- Environmentally Preferable:
Frequently Bought Together