Order Entry
United Kingdom
ContactUsLinkComponent
 
Platinum standard solution, 1000 mg/L Pt in 7% hydrochloric acid (from H₂PtCl₆) Certipur® standard for ICP, Supelco®
  1.70341.0100
 :  Merck
 :  
undefined
Platinum standard solution, 1000 mg/L Pt in 7% hydrochloric acid (from H₂PtCl₆) Certipur® standard for ICP, Supelco®
  1.70341.0100
 :  Merck
 :  

 

  • Concentration:
    1000 mg/L
  • Matrix:
    7% HCl
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Environmentally Preferable:
  • M-Clarity quality level:
    MQ100
  • Element:
    Platinum
  • Symbol:
    Pt

 

 

 : Traceable to SRM from NIST