Silicone oil for oil baths –40…+200°C
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Polyphenyl-methylsiloxane
CAS Number:63148-62-9
MDL Number:MFCD00132673
Density:0.963
Flash Pt:>270 °C (518 °F)
Merck Index:14,8495
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together