Reaction vessels, cylindrical, with flat flange
: KGW
Borosilicate glass.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
With thermal jacket, DN15 flange and drain valve.
Frequently Bought Together