Order Entry
United Kingdom
ContactUsLinkComponent
Tris-Acetate-EDTA (TAE) buffer, 50X
  A1691.1000
 :  
Tris-Acetate-EDTA (TAE) buffer, 50X
  A1691.1000
 :  
  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Description:
    Tris-Acetate-EDTA (TAE) buffer, 50X
  • Environmentally Preferable:
50X concentrated solution of 2 M Tris, 1 M glacial acetic acid and 0,5 M EDTA.Na₂, pH 8,5 ±0,2.