Dip slides for surfaces and in liquids, Envirocheck®
Supplier: Merck
Certificates
About this item
Envirocheck® contact slides are widely used to check critical control points (CCP's) within the HACCP (Hazardous Analysis Critical Control Point) for the microbiological hygiene control of surfaces in production lines and equipment. Additionally these slides can be used to check the microbiological hygiene status of liquids. The use of a variety of media on the contact slides allows the detection of organisms from liquids as well as surfaces. These slides provide semi-quantitative results for five different parameters. Easy incubation in the Cultura® mini incubator also reduces the necessary workspace. Each contact slide is packed in a PC tube, which protects the agar media against contamination before and after sampling.
- Flexible contact slide allows sampling even in difficult to reach areas
- Easy to use
- Protected against contamination
- Storage at <15 °C (ideally at 12 to 15 °C)
- Expiry date is written on each box
Packaging: 10 slides/box
Specifications
- Environmentally Preferable:
Specifications