Reagent kits for protein and fibre analysis
Catalog # 1.12979.0001
Supplier: Merck
CAS Number:
Specifications
- Pk:100 Tests
- Description:Total dietary fibre, enzymatic/gravimetric method
- Environmentally Preferable:
- Test method:Enzymatic-gravimetric
- Tests:100
Specifications
About this item
Simple reagent kits, used in food analysis, that use colour chemistry to determine levels of protein and fibre. Please note these kits require the use of a photometer or spectrophotometer.