Order Entry
United Kingdom
ContactUsLinkComponent
Gel for treatment of hydrofluoric acid burns, H-F Antidote
Gel for treatment of hydrofluoric acid burns, H-F Antidote
  118-0901
undefined
Gel for treatment of hydrofluoric acid burns, H-F Antidote
  118-0901
  • Description:
    Hydrofluoric acid burns gel, tube
  • Environmentally Preferable:
  • Pk:
    25 g
An emergency first aid treatment for hydrofluoric acid burns. If applied immediately the calcium gluconate in the gel combines with the hydrofluoric acid to form insoluble calcium fluoride preventing the extraction of calcium from bone and body tissue.
  • Significantly reduces damage caused by HF burns
  • Store above 8 °C and below 30 °C
Weight: 25 g