Stirrer shafts, with four bladed propeller, BOLA
Stirrer shafts made of PTFE-jacketed stainless steel and the propeller completely made of PTFE.
- With 45° angled angular blades
- Chemical resistant
- Temperature resistance from –200 to +250 °C
Frequently Bought Together