Accessories for JB Academy, JB Nova, SUB Aqua Pro, SUB Aqua Plus, JB Aqua Plus and SBB Aqua Plus baths
A range of different stainless steel accessories for use with JB Academy, JB Nova, SUB Aqua Pro, (also previous models: SUB Aqua Plus, JB Aqua Plus and SBB Aqua Plus baths).
- Flat lids with ring sets allow vessels of various sizes and heights to be used in the bath
- Sloping lids, with insulated handle, direct condensation down the side of the bath, protecting vessels in the bath
- Clear PC lids are supplied as standard with JB Nova and SUB Aqua Pro baths
- Raised half shelves help accommodate different sizes of vessel simultaneously by altering the effective liquid depth
- Wide range of test tube racks which are perforated to aid convection
Frequently Bought Together