Disinfectants, virucidal, Rely+On™ Virkon®
Supplier: DAY IMPEX
Certificates
About this item
Any viral disease transmitted via surfaces or instruments contaminated with infected blood, body fluids, faeces or saliva, can be controlled by the correct application of Virkon®. Independently tested and proven to inactivate viruses thus demonstrating its total and non-selective activity against them. Diluted in water to 1%, Virkon® can be used in spray or liquid form to clean and disinfect work surfaces and equipment in a single, one-step operation. A 50 g sachet can be diluted into 5 litres of solution, depending upon application.
- Widest proven spectrum of any disinfectant, excellent bactericide and fungicide
- Effective against all 22 virus families affecting people and animals
- In powder form can be directly applied to spillages of blood and other body fluids
- Odourless, 1% solution is non-irritant with exceptionally low oral toxicity
- Non corrosive, non bleaching
Ordering information: Available in 50×50 g sachets or 5 g tablets (can be divided between different laboratories), economical 1×5 kg drum, or 6×500 g squeeze ‘puff' packs.
Caution: Use biocides safely. Always read the label and product information before use.