Hazardous spill kits
: 3M
These absorption kits can be deployed wherever hazardous liquids are in use and can be used to contain and clean up a spill without delay.
- P110: For smaller spills or final clear-up of larger ones, sheets of 280×330 mm, absorption of 0,25 litre/sheet
- P130: Roll for large areas, can be cut to size, 33×3000 cm, absorption 50 litre/roll
- P190: Universal roll, 48×3300 cm, absorption 72 litre/roll
- P200: Minibooms, 75×1200 mm (Ø×l), absorption of 3,75 litre/miniboom
- P300: Pillows of 180×380 mm, absorption of 2 litre/pillow
- P-F2001: Folded sorbent that can be used as mini-booms, pillows, sheets or rolls, 12,5×1520 cm, perforated every 40,6 cm, absorption 119 litres/pack
Frequently Bought Together