Lotion soaps, antimicrobial, LabGuard™
Supplier: DAY IMPEX
Certificates
About this item
This antimicrobial hand soap contains 2% Nonoxynol 9, a concentrated coconut based lotion, safe enough to use as a shower soap. It also contains 0,5% p-Chloro-m-xylenol (PCMX) that is effective in destroying a broad spectrum of organisms including: Candida albicans, Rhizopus nigricans, Pseudomonas aeruginosa, Aerobacter/airogenus, Proteus vulgaris, Staphylococcus aureus, E.coli, B.cereus, B.subtilis, Streptococcus faecalis, Penicillium chrysogenum, Mocur recemosa, Altenaria solani, Oidium lactis, Aspergilius niger, Saccharomyces pastorianus, Torula roseus.
- Natural moisturisers and conditioners act gently but effectively on the skin, to combat the effects of water, harsh solutions and other irritants
- Guards against a wide range of bacterial and fungal organisms
- May be used repeatedly without drying the skin
Frequently Bought Together