2,4,6-Trimethylbenzyl chloride 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: α-2-Chloroisodurene, α-Chloro-1,2,4,6-tetramethylbenzene, α2-Chloroisodurene, alpha-2-Chloroisodurene, 1-(Chloromethyl)-2,4,6-trimethylbenzene
CAS Number:1585-16-6
:
Formula:C₁₀H₁₃Cl
MDL Number:MFCD00000901
Boiling Pt:114…115 °C (10 mmHg)
Molecular Weight:168.67 g/mol
Melting Pt:34…38 °C
Flash Pt:106 °C
Storage Temperature:Refrigerator
EINECS:216-440-3
UN:3261
ADR:8 II
:GHS05
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White Fused solid or crystals and/or chunks
- Infrared spectrum:Conforms
- Melting point:37 to 42 °C
- GC:≥97.5 %
- Residue after evaporation:≤0.0005 %
- Acidity (CH3COOH):≤0.002 %
Specification Test Results
Frequently Bought Together