Order Entry
United Kingdom
ContactUsLinkComponent
Fraction collectors, Foxy R1 and R2
Fraction collectors, Foxy R1 and R2
Certificates
undefined
Fraction collectors, Foxy R1 and R2
Specifications
  • Catalog No:
  • 905-0119
  • 905-0533
  • Model:
  • Foxy R1
  • Foxy R1
  • Max. flow rate:
  • 100 ml/min
  • 100 ml/min
  • Switch valves:
  • Diverter valve (3-way)
  • Diverter valve (3-way)
  • W×D×H:
  • 311×330×355 mm
  • 311×330×355 mm
  • Weight:
  • 7,1 kg
  • 7,1 kg
Specifications
Racks
Product Information
Frequently Bought Together