Bis(2-ethylhexyl)phosphate for synthesis
Supplier: Merck
Certificates
About this item
Synonyms: Bis(2-ethylhexyl) hydrogen phosphate, Di-(2-ethylhexyl)phosphoric acid
CAS Number:298-07-7
:
Formula:C₁₆H₃₅O₄P
MDL Number:MFCD00009492
Molecular Weight:322.43 g/mol
ADR:8 III
:M-Clarity™ quality level = MQ200
EINECS:206-056-4
UN:1902
:GHS05
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Solubility:- 1 g/l (20 °C)
- Melting Point:-50 °C
- Molar Mass:322.42 g/mol
- Boiling Point:48 °C (16 hPa)
- Vapor Pressure:- 0.1 hPa (20 °C)
- Flash Point:150 °C
- Refractive Index:1.443 (20 °C, 589 nm)
- Density:0.97 g/cm3 (20 °C)
- pH:3 ( 1 g/l, H2O)
- Ignition Point:300 °C
- Synonyms:HDEHP, Phosphoric acid bis (2-ethylhexyl) ester, Bis(2-ethylhexyl) phosphoric acid
- Hill Formula:C16H35O4P
- Chemical Formula:(CH3(CH2)3CH(C2H5)CH2O)2POOH
Specification Test Results
Frequently Bought Together